Product Details

SNP ID
rs204910
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44960128 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCTGTCTGATTTCCCTTCCCCAGC[C/T]GACCCCCGCCCCATAGATCTCCTCA
Phenotype
MIM: 604783
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CLPTM1 PubMed Links
Additional Information
For this assay, SNP(s) [rs116949436] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CLPTM1
Gene Name
CLPTM1, transmembrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282175.1 Intron NP_001269104.1
NM_001282176.1 Intron NP_001269105.1
NM_001294.3 Intron NP_001285.1

View Full Product Details