Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_152505.3 | Intron | NP_689718.1 | |||
XM_005260926.1 | Intron | XP_005260983.1 | |||
XM_006723967.1 | Intron | XP_006724030.1 | |||
XM_006723972.1 | Intron | XP_006724035.1 | |||
XM_006723973.1 | Intron | XP_006724036.1 | |||
XM_011529458.1 | Intron | XP_011527760.1 | |||
XM_011529459.1 | Intron | XP_011527761.1 | |||
XM_011529460.1 | Intron | XP_011527762.1 | |||
XM_011529461.1 | Intron | XP_011527763.1 | |||
XM_011529462.1 | Intron | XP_011527764.1 | |||
XM_011529463.2 | Intron | XP_011527765.1 | |||
XM_011529464.1 | Intron | XP_011527766.1 | |||
XM_011529465.1 | Intron | XP_011527767.1 | |||
XM_011529466.1 | Intron | XP_011527768.1 | |||
XM_011529467.1 | Intron | XP_011527769.1 | |||
XM_011529468.1 | Intron | XP_011527770.1 | |||
XM_011529469.2 | Intron | XP_011527771.1 | |||
XM_011529470.1 | Intron | XP_011527772.1 | |||
XM_011529471.1 | Intron | XP_011527773.1 | |||
XM_017028274.1 | Intron | XP_016883763.1 | |||
XM_017028275.1 | Intron | XP_016883764.1 | |||
XM_017028276.1 | Intron | XP_016883765.1 | |||
XM_017028277.1 | Intron | XP_016883766.1 | |||
XM_017028278.1 | Intron | XP_016883767.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001001713.1 | Intron | NP_001001713.1 | |||
NM_001317740.1 | Intron | NP_001304669.1 | |||
NM_001317741.1 | Intron | NP_001304670.1 | |||
NM_001317742.1 | Intron | NP_001304671.1 | |||
NM_007341.2 | Intron | NP_031367.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001317744.1 | Intron | NP_001304673.1 |