Product Details

SNP ID
rs974603
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.21:28573437 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAATAGAATCTTGTAGTAAAACTT[A/G]TAATTTGTCACAATTGTAAATCAAA
Phenotype
MIM: 614553
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
N6AMT1 PubMed Links
Additional Information
For this assay, SNP(s) [rs113479615] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
N6AMT1
Gene Name
N-6 adenine-specific DNA methyltransferase 1 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013240.5 Intron NP_037372.4
NM_182749.4 Intron NP_877426.4

View Full Product Details