Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001037161.1 | Intron | NP_001032238.1 | |||
XM_017021589.1 | Intron | XP_016877078.1 | |||
XM_017021590.1 | Intron | XP_016877079.1 | |||
XM_017021591.1 | Intron | XP_016877080.1 | |||
XM_017021592.1 | Intron | XP_016877081.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001220484.1 | Intron | NP_001207413.1 | |||
NM_203309.2 | Intron | NP_976054.2 | |||
XM_006720143.2 | Intron | XP_006720206.1 | |||
XM_011536760.2 | Intron | XP_011535062.1 | |||
XM_011536761.2 | Intron | XP_011535063.1 | |||
XM_017021289.1 | Intron | XP_016876778.1 |