Product Details

SNP ID
rs3782422
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:113821920 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCTTGCCTGGGCTCTGGGCCACTC[A/C]GAAAACTGGATGAAGACTGGAACCT
Phenotype
MIM: 616444
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RBM19 PubMed Links

Gene Details

Gene
RBM19
Gene Name
RNA binding motif protein 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146698.1 Intron NP_001140170.1
NM_001146699.1 Intron NP_001140171.1
NM_016196.3 Intron NP_057280.2
XM_017020280.1 Intron XP_016875769.1
XM_017020281.1 Intron XP_016875770.1

View Full Product Details