Product Details

SNP ID
rs2145455
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:105841492 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCAGACTCGGGCCTTCGTTGCTTC[C/T]AGAGCTCTCTGCTCTGTGGCTGCCA
Phenotype
MIM: 146910 MIM: 147070 MIM: 147010 MIM: 147170
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGH PubMed Links

Gene Details

Gene
IGH
Gene Name
immunoglobulin heavy locus
There are no transcripts associated with this gene.

Gene
IGHD
Gene Name
immunoglobulin heavy constant delta
There are no transcripts associated with this gene.

View Full Product Details