Product Details

SNP ID
rs852964
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:71413926 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATGTGGTTTTTTATTTTTCTATTA[A/G]AATTTAGGAATAAGTTTAACTATTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00472 PubMed Links

Gene Details

Gene
LINC00472
Gene Name
long intergenic non-protein coding RNA 472
There are no transcripts associated with this gene.

Gene
LINC01626
Gene Name
long intergenic non-protein coding RNA 1626
There are no transcripts associated with this gene.

View Full Product Details