Product Details

SNP ID
rs977747
Assay Type
Functionally tested
NCBI dbSNP Submissions
55
Location
Chr.1:47219005 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGCTTTCTCAGGGCTGCAGACAT[G/T]GTCTCCACAGCCACAGGCTTAGGAA
Phenotype
MIM: 187040
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TAL1 PubMed Links

Gene Details

Gene
TAL1
Gene Name
TAL bHLH transcription factor 1, erythroid differentiation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287347.2 2037 UTR 3 NP_001274276.1
NM_001290403.1 2037 UTR 3 NP_001277332.1
NM_001290404.1 2037 UTR 3 NP_001277333.1
NM_001290405.1 2037 UTR 3 NP_001277334.1
NM_001290406.1 2037 UTR 3 NP_001277335.1
NM_003189.5 2037 UTR 3 NP_003180.1
XM_005271160.4 2037 UTR 3 XP_005271217.1
XM_017002187.1 2037 UTR 3 XP_016857676.1
XM_017002188.1 2037 UTR 3 XP_016857677.1
XM_017002189.1 2037 UTR 3 XP_016857678.1
XM_017002190.1 2037 UTR 3 XP_016857679.1
XM_017002191.1 2037 UTR 3 XP_016857680.1
XM_017002192.1 2037 Intron XP_016857681.1
XM_017002193.1 2037 UTR 3 XP_016857682.1

View Full Product Details