Product Details

SNP ID
rs35343305
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:101356571 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
ACATTATTAAATCTTGAATCAGTTT[C/T]CATAGGGAGAAGCCAATTTTTTCTT
Phenotype
MIM: 603482
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BTRC PubMed Links

Gene Details

Gene
BTRC
Gene Name
beta-transducin repeat containing E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256856.1 Intron NP_001243785.1
NM_003939.4 Intron NP_003930.1
NM_033637.3 Intron NP_378663.1
XM_006718054.2 Intron XP_006718117.1
XM_011540320.2 Intron XP_011538622.1
XM_017016870.1 Intron XP_016872359.1
XM_017016871.1 Intron XP_016872360.1
XM_017016872.1 Intron XP_016872361.1
XM_017016873.1 Intron XP_016872362.1
XM_017016874.1 Intron XP_016872363.1

View Full Product Details