Product Details

SNP ID
rs2823597
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:16075052 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGGAAATGATATCTGGGTTCACT[C/T]TGCTAGCTCTGAATTCAGTTAACAC
Phenotype
MIM: 615964
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR99AHG PubMed Links

Gene Details

Gene
MIR99AHG
Gene Name
mir-99a-let-7c cluster host gene
There are no transcripts associated with this gene.

View Full Product Details