Product Details

SNP ID
rs1344709
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:44572256 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AACTCTACTCTATCAGGCCAAAATA[C/T]AGGCAACTACATGTAACGTACCCGT
Phenotype
MIM: 600623
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD82 PubMed Links

Gene Details

Gene
CD82
Gene Name
CD82 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024844.1 Intron NP_001020015.1
NM_002231.3 Intron NP_002222.1
XM_006718223.2 Intron XP_006718286.1
XM_011520067.2 Intron XP_011518369.1

View Full Product Details