Product Details

SNP ID
rs26524
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:103120600 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CCTGTGCTCTGCAGACCCTAAACAC[A/G]TGCTCCACCGAGGGGCTTGTGCTGG
Phenotype
MIM: 611648
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GIN1 PubMed Links

Gene Details

Gene
GIN1
Gene Name
gypsy retrotransposon integrase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317954.1 Intron NP_001304883.1
NM_017676.2 Intron NP_060146.2
XM_005272028.3 Intron XP_005272085.1
XM_017009599.1 Intron XP_016865088.1
Gene
PPIP5K2
Gene Name
diphosphoinositol pentakisphosphate kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276277.1 Intron NP_001263206.1
NM_001281471.1 Intron NP_001268400.1
NM_015216.3 Intron NP_056031.2
XM_005271935.1 Intron XP_005271992.1
XM_005271936.1 Intron XP_005271993.1
XM_005271938.1 Intron XP_005271995.1
XM_005277534.1 Intron XP_005277591.1
XM_005277535.1 Intron XP_005277592.1
XM_005277536.1 Intron XP_005277593.1
XM_005277538.1 Intron XP_005277595.1
XM_005277539.1 Intron XP_005277596.1
XM_005277541.1 Intron XP_005277598.1
XM_006714576.1 Intron XP_006714639.1
XM_006714577.1 Intron XP_006714640.1
XM_011543290.2 Intron XP_011541592.1
XM_017009269.1 Intron XP_016864758.1
XM_017009270.1 Intron XP_016864759.1
XM_017009271.1 Intron XP_016864760.1
XM_017009272.1 Intron XP_016864761.1
XM_017009273.1 Intron XP_016864762.1
XM_017009274.1 Intron XP_016864763.1
XM_017009275.1 Intron XP_016864764.1

View Full Product Details