Product Details

SNP ID
rs11864839
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31083930 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGCCATGACAGATGAGAATACTGA[C/G]GCTCAAAGCGGTTGAGCAGCCTGCT
Phenotype
MIM: 610561 MIM: 608547
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRSS53 PubMed Links

Gene Details

Gene
PRSS53
Gene Name
protease, serine 53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039503.2 3057 Intron NP_001034592.1
XM_011545816.1 3057 Intron XP_011544118.1
XM_011545817.1 3057 UTR 3 XP_011544119.1
XM_011545818.2 3057 Intron XP_011544120.1
XM_011545819.1 3057 Intron XP_011544121.1
XM_011545820.1 3057 UTR 3 XP_011544122.1
Gene
VKORC1
Gene Name
vitamin K epoxide reductase complex subunit 1
There are no transcripts associated with this gene.

Gene
ZNF646
Gene Name
zinc finger protein 646
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014699.3 3057 Intron NP_055514.3
XM_005255710.3 3057 Intron XP_005255767.1
XM_005255711.3 3057 Intron XP_005255768.1
XM_005255712.3 3057 Intron XP_005255769.1
XM_011545990.2 3057 Intron XP_011544292.1
Gene
ZNF668
Gene Name
zinc finger protein 668
There are no transcripts associated with this gene.

View Full Product Details