Product Details

SNP ID
rs11642526
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:71738424 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAACTAAAATTTGGTGTAAACCAA[A/T]CCTTGATTCACAAAAACAGTTACTG
Phenotype
MIM: 603533
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
AP1G1 PubMed Links

Gene Details

Gene
AP1G1
Gene Name
adaptor related protein complex 1 gamma 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001030007.1 Intron NP_001025178.1
NM_001128.5 Intron NP_001119.3

View Full Product Details