Product Details

SNP ID
rs7197664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:79204463 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGCATGTCTAGGGTCTCACAATAA[C/T]AGCAGGTCTCAGAAAGATAAAATAA
Phenotype
MIM: 177075 MIM: 605131
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105371352 PubMed Links

Gene Details

Gene
LOC105371352
Gene Name
uncharacterized LOC105371352
There are no transcripts associated with this gene.

Gene
MAF
Gene Name
MAF bZIP transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031804.2 Intron NP_001026974.1
NM_005360.4 Intron NP_005351.2
XM_017023233.1 Intron XP_016878722.1
XM_017023234.1 Intron XP_016878723.1
XM_017023235.1 Intron XP_016878724.1
Gene
WWOX
Gene Name
WW domain containing oxidoreductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291997.1 Intron NP_001278926.1
NM_016373.3 Intron NP_057457.1
NM_130791.3 Intron NP_570607.1
XM_011523101.2 Intron XP_011521403.1
XM_011523103.2 Intron XP_011521405.1
XM_011523104.2 Intron XP_011521406.1
XM_011523105.2 Intron XP_011521407.1
XM_017023278.1 Intron XP_016878767.1
XM_017023279.1 Intron XP_016878768.1

View Full Product Details