Product Details

SNP ID
rs1014064
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:147854585 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TACTGACTCTTCTTTGACCTTTGAC[A/G]GCCATTAAGATTTCACTTTACTCAC
Phenotype
MIM: 102581
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACVR2A PubMed Links

Gene Details

Gene
ACVR2A
Gene Name
activin A receptor type 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278579.1 Intron NP_001265508.1
NM_001278580.1 Intron NP_001265509.1
NM_001616.4 Intron NP_001607.1

View Full Product Details