Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001099433.1 | Intron | NP_001092903.1 | |||
NM_001306133.1 | Intron | NP_001293062.1 | |||
NM_001306134.1 | Intron | NP_001293063.1 | |||
NM_144720.3 | Intron | NP_653321.1 | |||
XM_011513400.2 | Intron | XP_011511702.1 | |||
XM_017007788.1 | Intron | XP_016863277.1 | |||
XM_017007789.1 | Intron | XP_016863278.1 | |||
XM_017007790.1 | Intron | XP_016863279.1 | |||
XM_017007791.1 | Intron | XP_016863280.1 | |||
XM_017007792.1 | Intron | XP_016863281.1 | |||
XM_017007793.1 | Intron | XP_016863282.1 | |||
XM_017007794.1 | Intron | XP_016863283.1 | |||
XM_017007795.1 | Intron | XP_016863284.1 | |||
XM_017007796.1 | Intron | XP_016863285.1 | |||
XM_017007797.1 | Intron | XP_016863286.1 | |||
XM_017007798.1 | Intron | XP_016863287.1 | |||
XM_017007799.1 | Intron | XP_016863288.1 |