Product Details

SNP ID
rs12451505
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7722570 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCACTCCTGATTCCCCCTTCCCCC[C/G]CAGATCCCAGGAACCGCTCATCTAT
Phenotype
MIM: 603333
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNAH2 PubMed Links

Gene Details

Gene
DNAH2
Gene Name
dynein axonemal heavy chain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303270.1 Intron NP_001290199.1
NM_020877.3 Intron NP_065928.2
XM_011523663.1 Intron XP_011521965.1
XM_011523664.2 Intron XP_011521966.1
XM_011523666.1 Intron XP_011521968.1
XM_011523667.2 Intron XP_011521969.1
XM_011523668.1 Intron XP_011521970.1
XM_011523669.2 Intron XP_011521971.1
XM_011523670.2 Intron XP_011521972.1
XM_017024218.1 Intron XP_016879707.1
XM_017024219.1 Intron XP_016879708.1

View Full Product Details