Product Details

SNP ID
rs747528
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:14506507 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTTACACAGTGAAAAATGGATGGGA[A/G]AGAGAACCAGATAAACGTCCATCAG
Phenotype
MIM: 613644
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATF7IP PubMed Links

Gene Details

Gene
ATF7IP
Gene Name
activating transcription factor 7 interacting protein
There are no transcripts associated with this gene.

Gene
PLBD1
Gene Name
phospholipase B domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024829.5 Intron NP_079105.4

View Full Product Details