Product Details
- SNP ID
-
rs10783394
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:50846895 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATCCGGCCTGCAGTAAGTTTTGAAA[C/T]CAGGAAATGTGAGCCCTCCAACTTT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TMPRSS12
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs12317842,rs533581364] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TMPRSS12
- Gene Name
- transmembrane protease, serine 12
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_182559.2 |
|
Intron |
|
|
NP_872365.1 |
View Full Product Details