Product Details

SNP ID
rs334996
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:29924412 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGTTTCAAGGACAGGAGCCCCCCA[C/T]TGTTGCTCGGTGTACTGGCGATCTA
Phenotype
MIM: 603494
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
URI1 PubMed Links

Gene Details

Gene
URI1
Gene Name
URI1, prefoldin like chaperone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252641.1 Intron NP_001239570.1
NM_003796.3 Intron NP_003787.2
XM_005259362.2 Intron XP_005259419.1
XM_005259363.4 Intron XP_005259420.1
XM_011527435.1 Intron XP_011525737.1

View Full Product Details