Product Details

SNP ID
rs7255306
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51333917 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGTGACCACGGGGACCAAGGTAGA[A/G]GGATGCTTCTTTTTCTCAGGAGTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IGLON5 PubMed Links

Gene Details

Gene
IGLON5
Gene Name
IgLON family member 5
There are no transcripts associated with this gene.

Gene
VSIG10L
Gene Name
V-set and immunoglobulin domain containing 10 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163922.1 2448 Silent Mutation CCC,CCT P816P NP_001157394.1

View Full Product Details