Product Details

SNP ID
rs773070
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:42902717 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACCCTTCTATAGGCTATTTTTCCC[C/G]AATCTTTCTTTATATGGAAAAACTG
Phenotype
MIM: 607825 MIM: 614649
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HOOK3 PubMed Links

Gene Details

Gene
HOOK3
Gene Name
hook microtubule-tethering protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032410.3 Intron NP_115786.1
Gene
MIR4469
Gene Name
microRNA 4469
There are no transcripts associated with this gene.

Gene
RNF170
Gene Name
ring finger protein 170
There are no transcripts associated with this gene.

View Full Product Details