Product Details
- SNP ID
-
rs2009192
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:139940740 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- GGCTTGACTTGTTTGTAAAAAGACA[C/T]GCTTTTTTTCACTAGTCTATGGATG
- Phenotype
-
MIM: 611323
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CLSTN2
PubMed Links
Gene Details
- Gene
- CLSTN2
- Gene Name
- calsyntenin 2
View Full Product Details