Product Details

SNP ID
rs11038866
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:46334382 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCACTAGGGCAGCTGCGGTCCTTA[C/G]AGCCATCGGCCTGGTTGGAGAGGAC
Phenotype
MIM: 601441
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DGKZ PubMed Links

Gene Details

Gene
DGKZ
Gene Name
diacylglycerol kinase zeta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105540.1 Intron NP_001099010.1
NM_001199266.1 Intron NP_001186195.1
NM_001199267.1 Intron NP_001186196.1
NM_001199268.1 Intron NP_001186197.1
NM_003646.3 Intron NP_003637.2
NM_201532.2 Intron NP_963290.1
NM_201533.3 Intron NP_963291.2
XM_005253181.3 Intron XP_005253238.1
XM_005253182.3 Intron XP_005253239.1
XM_006718354.1 Intron XP_006718417.1
XM_006718355.2 Intron XP_006718418.1
XM_011520422.1 Intron XP_011518724.1
XM_011520423.1 Intron XP_011518725.1
XM_017018455.1 Intron XP_016873944.1
XM_017018456.1 Intron XP_016873945.1
XM_017018457.1 Intron XP_016873946.1

View Full Product Details