Product Details

SNP ID
rs12606984
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:74437618 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGACAAAAACCTTCAACTTCCATTT[A/C]ACAATCCCTGCAAAAGCAAGGATTC
Phenotype
MIM: 614544
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM69C PubMed Links

Gene Details

Gene
FAM69C
Gene Name
family with sequence similarity 69 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001044369.2 Intron NP_001037834.2
XM_011525816.2 Intron XP_011524118.1
XM_017025551.1 Intron XP_016881040.1

View Full Product Details