Product Details

SNP ID
rs1820518
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:45402381 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTCGGTGTTCCGCTTTTTGTGGGC[C/T]GGGTGGGTTTCCTAATCTGGTTTCG
Phenotype
MIM: 602360
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GATM PubMed Links

Gene Details

Gene
GATM
Gene Name
glycine amidinotransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321015.1 61 Intron NP_001307944.1
NM_001482.2 61 Intron NP_001473.1
Gene
SPATA5L1
Gene Name
spermatogenesis associated 5 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323640.1 61 UTR 5 NP_001310569.1
NM_024063.2 61 UTR 5 NP_076968.2

View Full Product Details