Product Details

SNP ID
rs12207807
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:170538410 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGTATGTTTTCTACTATAGAACTG[C/T]GTAAACTTGGCCAGGCGCGGTGGCT
Phenotype
MIM: 602017
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSMB1 PubMed Links

Gene Details

Gene
PSMB1
Gene Name
proteasome subunit beta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002793.3 Intron NP_002784.1

View Full Product Details