Product Details

SNP ID
rs270200
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:5141372 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGGGCCACAATTTGACATGAGG[A/G]CAAGTCTTATTCTTTTTCAATGCAT
Phenotype
MIM: 607510
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAMTS16 PubMed Links

Gene Details

Gene
ADAMTS16
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139056.3 Intron NP_620687.2
Gene
CTD-2297D10.2
Gene Name
uncharacterized LOC101929176
There are no transcripts associated with this gene.

View Full Product Details