Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001317948.1 | Intron | NP_001304877.1 | |||
NM_173664.5 | Intron | NP_775935.1 | |||
XM_011534529.2 | Intron | XP_011532831.1 | |||
XM_011534530.2 | Intron | XP_011532832.1 | |||
XM_011534531.2 | Intron | XP_011532833.1 | |||
XM_017009372.1 | Intron | XP_016864861.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001834.3 | Intron | NP_001825.1 | |||
NM_007097.3 | Intron | NP_009028.1 | |||
XM_017009026.1 | Intron | XP_016864515.1 | |||
XM_017009027.1 | Intron | XP_016864516.1 |