Product Details

SNP ID
rs2241581
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:178241881 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACCAGAGCAAGGGGAACCGCGATGA[C/T]GGCAGTAGCAGCAGCAGTGGCCCTG
Phenotype
MIM: 610043 MIM: 614683
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COL23A1 PubMed Links

Gene Details

Gene
COL23A1
Gene Name
collagen type XXIII alpha 1 chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173465.3 Intron NP_775736.2
XM_006714933.3 Intron XP_006714996.1
XM_006714934.3 Intron XP_006714997.1
XM_006714935.3 Intron XP_006714998.1
XM_006714936.3 Intron XP_006714999.1
XM_011534688.2 Intron XP_011532990.1
XM_011534689.2 Intron XP_011532991.1
XM_011534690.2 Intron XP_011532992.1
XM_011534691.2 Intron XP_011532993.1
XM_011534692.2 Intron XP_011532994.1
XM_017010018.1 Intron XP_016865507.1
Gene
PHYKPL
Gene Name
5-phosphohydroxy-L-lysine phospho-lyase
There are no transcripts associated with this gene.

View Full Product Details