Product Details
- SNP ID
-
rs2033218
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:24611691 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTCAATGTAATTGCAGTGGAGATGG[A/T]CAGGGAGAGGACAGATTAGAATGAT
- Phenotype
-
MIM: 610739
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TNRC6A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113075928] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TNRC6A
- Gene Name
- trinucleotide repeat containing 6A
View Full Product Details