Product Details

SNP ID
rs12983784
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:7960607 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CACAAATTCGGAGCTGCCTGGGGTC[C/T]AACTACCAGAGGAAGTTACACACGG
Phenotype
MIM: 603466
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ELAVL1 PubMed Links

Gene Details

Gene
ELAVL1
Gene Name
ELAV like RNA binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001419.2 4024 UTR 3 NP_001410.2

View Full Product Details