Product Details

SNP ID
rs277416
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:41926198 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCCAATGTGTATATCAACGTGAAA[A/G]CTGCAGGTTTAGAATGCCTCCAAGT
Phenotype
MIM: 609090
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C5orf51 PubMed Links

Gene Details

Gene
C5orf51
Gene Name
chromosome 5 open reading frame 51
There are no transcripts associated with this gene.

Gene
FBXO4
Gene Name
F-box protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297437.1 Intron NP_001284366.1
NM_012176.2 Intron NP_036308.1
NM_033484.2 Intron NP_277019.1
XM_006714470.3 Intron XP_006714533.1
XM_011514026.2 Intron XP_011512328.1
XM_011514027.2 Intron XP_011512329.1

View Full Product Details