Product Details

SNP ID
rs2163417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:30615081 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATGGAAAAATGAAGTGTATACCT[C/T]AAGTGATACAGTCTTGGCTGAATCA
Phenotype
MIM: 190182
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TGFBR2 PubMed Links

Gene Details

Gene
TGFBR2
Gene Name
transforming growth factor beta receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024847.2 Intron NP_001020018.1
NM_003242.5 Intron NP_003233.4
XM_011534043.2 Intron XP_011532345.1
XM_011534045.2 Intron XP_011532347.1
XM_017007106.1 Intron XP_016862595.1

View Full Product Details