Product Details

SNP ID
rs34411705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:82355233 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGAAACTCTTAAATTAAGCATT[A/G]TTTTATGAACAATATAGATTATAAA
Phenotype
MIM: 601324
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HNRNPD PubMed Links

Gene Details

Gene
HNRNPD
Gene Name
heterogeneous nuclear ribonucleoprotein D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003810.1 Intron NP_001003810.1
NM_002138.3 Intron NP_002129.2
NM_031369.2 Intron NP_112737.1
NM_031370.2 Intron NP_112738.1

View Full Product Details