Product Details
- SNP ID
-
rs11827585
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:117820660 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCCGCACCTTCCCCAGGCCCTCCTA[G/T]CATACCTGCTGTTACGGCTCATCTT
- Phenotype
-
MIM: 601814
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FXYD2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs118032332] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FXYD2
- Gene Name
- FXYD domain containing ion transport regulator 2
- Gene
- FXYD6-FXYD2
- Gene Name
- FXYD6-FXYD2 readthrough
View Full Product Details