Product Details

SNP ID
rs2013182
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:58560409 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTCCGCCCCCGTTCAAATGCCCA[A/G]GACAGGGAATCATCCAGCCACTGCC
Phenotype
MIM: 605792
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEPT4-AS1 PubMed Links

Gene Details

Gene
SEPT4-AS1
Gene Name
SEPT4 antisense RNA 1
There are no transcripts associated with this gene.

Gene
TEX14
Gene Name
testis expressed 14, intercellular bridge forming factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201457.1 Intron NP_001188386.1
NM_031272.4 Intron NP_112562.3
NM_198393.3 Intron NP_938207.2
XM_011525029.2 Intron XP_011523331.1
XM_017024860.1 Intron XP_016880349.1
XM_017024861.1 Intron XP_016880350.1

View Full Product Details