Product Details

SNP ID
rs4675212
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:201649160 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AATTGGGCCTACGGATTGCATGCTG[C/T]TTATCTTACCTTAACTTGCTCACCC
Phenotype
MIM: 606575 MIM: 614423
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MPP4 PubMed Links

Gene Details

Gene
MPP4
Gene Name
membrane palmitoylated protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033066.2 Intron NP_149055.1
XM_017004620.1 Intron XP_016860109.1
Gene
TMEM237
Gene Name
transmembrane protein 237
There are no transcripts associated with this gene.

View Full Product Details