Product Details

SNP ID
rs8120279
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3472116 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTCCCTGGAGGTGGCAAATTGGAA[C/T]AGGGTCTGTGGTGAAGAGGGAATGG
Phenotype
MIM: 603130
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATRN PubMed Links

Gene Details

Gene
ATRN
Gene Name
attractin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207047.2 Intron NP_001193976.1
NM_001323332.1 Intron NP_001310261.1
NM_139321.2 Intron NP_647537.1
NM_139322.3 Intron NP_647538.1

View Full Product Details