Product Details

SNP ID
rs243629
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.21:17848481 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGACTGGACCGCATGTCTCTATCAC[C/T]GAACAGTCCAGCCCAGGGTTGTTCA
Phenotype
MIM: 607247
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHODL PubMed Links

Gene Details

Gene
CHODL
Gene Name
chondrolectin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204174.1 Intron NP_001191103.1
NM_001204175.1 Intron NP_001191104.1
NM_001204176.1 Intron NP_001191105.1
NM_001204177.1 Intron NP_001191106.1
NM_001204178.1 Intron NP_001191107.1
NM_024944.2 Intron NP_079220.2
XM_011529453.2 Intron XP_011527755.1
XM_011529457.2 Intron XP_011527759.1
XM_017028273.1 Intron XP_016883762.1
Gene
CHODL-AS1
Gene Name
CHODL antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details