Product Details

SNP ID
rs11884190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:201118704 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTGGCGGGGATTAGGCGTGAAGC[A/G]GTTCAGCAGGCAGAGGTTCTCGGAC
Phenotype
MIM: 603599
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFLAR PubMed Links

Gene Details

Gene
CFLAR
Gene Name
CASP8 and FADD like apoptosis regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127183.2 2139 UTR 5 NP_001120655.1
NM_001127184.2 2139 Intron NP_001120656.1
NM_001202515.1 2139 Intron NP_001189444.1
NM_001202516.1 2139 Intron NP_001189445.1
NM_001202517.1 2139 Intron NP_001189446.1
NM_001202518.1 2139 Intron NP_001189447.1
NM_001202519.1 2139 Intron NP_001189448.1
NM_001308042.1 2139 Intron NP_001294971.1
NM_001308043.1 2139 Intron NP_001294972.1
NM_003879.5 2139 Intron NP_003870.4
XM_017005189.1 2139 Intron XP_016860678.1
XM_017005190.1 2139 Intron XP_016860679.1
XM_017005191.1 2139 Intron XP_016860680.1
XM_017005192.1 2139 Intron XP_016860681.1
XM_017005193.1 2139 UTR 5 XP_016860682.1
XM_017005194.1 2139 Intron XP_016860683.1
XM_017005195.1 2139 Intron XP_016860684.1
XM_017005196.1 2139 Intron XP_016860685.1
XM_017005197.1 2139 UTR 5 XP_016860686.1
XM_017005198.1 2139 Intron XP_016860687.1

View Full Product Details