Product Details

SNP ID
rs12879694
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:93186501 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTGGGTGTGGACAGTTATTTTT[A/C]TGGTACCTTACTTACAGATGTTTTT
Phenotype
MIM: 609485
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
MOAP1 PubMed Links

Gene Details

Gene
MOAP1
Gene Name
modulator of apoptosis 1
There are no transcripts associated with this gene.

Gene
TMEM251
Gene Name
transmembrane protein 251
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098621.1 698 Missense Mutation ATG,CTG M114L NP_001092091.1
NM_015676.1 698 Missense Mutation NP_056491.1

View Full Product Details