Product Details

SNP ID
rs738679
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.22:43415773 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GTAGGCTCCTCTTTCCACCACCCCA[A/G]TCTACACCTGAAAGTTTGTTTCTAT
Phenotype
MIM: 602112
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MPPED1 PubMed Links

Gene Details

Gene
MPPED1
Gene Name
metallophosphoesterase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001044370.1 3359 Intron NP_001037835.1
XM_011530367.2 3359 UTR 5 XP_011528669.1

View Full Product Details