Product Details

SNP ID
rs3209284
Assay Type
Validated
NCBI dbSNP Submissions
53
Location
Chr.1:11919852 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
ACCGTGGGGCCTCCGAACGCCGAGA[C/T]GGACATCACCGCCAACACTCACCAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA2013 PubMed Links

Gene Details

Gene
KIAA2013
Gene Name
KIAA2013
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138346.2 2559 UTR 3 NP_612355.1
XM_005263531.4 2559 Intron XP_005263588.1

View Full Product Details