Product Details

SNP ID
rs3746611
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.20:32445771 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TATTGGAGGCCACTTGAGAGATAAC[A/G]ACCACCCCCAACCTCTAGGGGCCGG
Phenotype
MIM: 612990
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASXL1 PubMed Links

Gene Details

Gene
ASXL1
Gene Name
additional sex combs like 1, transcriptional regulator
There are no transcripts associated with this gene.

Gene
NOL4L
Gene Name
nucleolar protein 4 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256798.1 3390 UTR 3 NP_001243727.1
NM_080616.4 3390 UTR 3 NP_542183.2
XM_005260282.4 3390 Intron XP_005260339.1
XM_005260283.4 3390 Intron XP_005260340.1
XM_005260284.4 3390 Intron XP_005260341.1
XM_005260285.4 3390 Intron XP_005260342.2
XM_005260286.4 3390 Intron XP_005260343.1
XM_005260288.2 3390 Intron XP_005260345.1
XM_005260289.4 3390 Intron XP_005260346.1
XM_006723702.3 3390 Intron XP_006723765.1
XM_006723703.3 3390 Intron XP_006723766.1
XM_011528562.2 3390 Intron XP_011526864.1
XM_011528563.1 3390 Intron XP_011526865.1
XM_017027669.1 3390 Intron XP_016883158.1
XM_017027670.1 3390 Intron XP_016883159.1

View Full Product Details