Product Details
- SNP ID
-
rs2425670
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:44893088 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTATTTAAAAAAAAATGGATTAGCA[A/G]TAGGTAGTCTTCTTTTTCTTTTCTT
- Phenotype
-
MIM: 601289
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
YWHAB
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79248642] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- YWHAB
- Gene Name
- tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein beta
View Full Product Details