Product Details

SNP ID
rs4930077
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:2298258 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCCAGAGAGAGGGCAGCCTGGCAA[C/T]GGGCCTGGGCCCTGACGCTTGAGCA
Phenotype
MIM: 611033 MIM: 603853
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C11orf21 PubMed Links

Gene Details

Gene
C11orf21
Gene Name
chromosome 11 open reading frame 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011520034.1 Intron XP_011518336.1
Gene
TSPAN32
Gene Name
tetraspanin 32
There are no transcripts associated with this gene.

View Full Product Details