Product Details

SNP ID
rs11865184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57667974 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTGTGAGAGGAGGGGTCTGAGTA[A/G]GTGCGCCTGACTCCAAAGCCTGTGC
Phenotype
MIM: 604110
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADGRG1 PubMed Links

Gene Details

Gene
ADGRG1
Gene Name
adhesion G protein-coupled receptor G1
There are no transcripts associated with this gene.

Gene
ADGRG3
Gene Name
adhesion G protein-coupled receptor G3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308360.1 Intron NP_001295289.1
NM_170776.4 Intron NP_740746.4
XM_005255842.3 Intron XP_005255899.1
XM_006721170.3 Intron XP_006721233.1
XM_011522951.2 Intron XP_011521253.1
XM_011522953.2 Intron XP_011521255.1
XM_011522954.1 Intron XP_011521256.1

View Full Product Details