Product Details

SNP ID
rs11177691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:69483513 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTACAAAAATAGAAAAAAATTCTTA[A/T]TTCTATCACTAATACAGTCAATTTT
Phenotype
MIM: 607743
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FRS2 PubMed Links

Gene Details

Gene
FRS2
Gene Name
fibroblast growth factor receptor substrate 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042555.2 Intron NP_001036020.1
NM_001278351.1 Intron NP_001265280.1
NM_001278353.1 Intron NP_001265282.1
NM_001278354.1 Intron NP_001265283.1
NM_001278355.1 Intron NP_001265284.1
NM_001278356.1 Intron NP_001265285.1
NM_001278357.1 Intron NP_001265286.1
NM_006654.4 Intron NP_006645.3
XM_017018717.1 Intron XP_016874206.1
XM_017018718.1 Intron XP_016874207.1
XM_017018719.1 Intron XP_016874208.1

View Full Product Details